Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK3B Polyclonal Antibody, Invitrogen™

Catalog No. PIPA595845
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595845 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595845 Supplier Thermo Scientific Supplier No. PA595845

Rabbit Polyclonal Antibody

Immunogen sequence: LPNGRDTPAL FNFTTQELSS NPPLATILIP PHARIQAAAS TPTNATAASD ANTGDRGQTN NAASASASNS T

GSK3B is a proline-directed serine/threonine protein kinase involved in energy metabolism, neuronal cell development and body pattern formation. It is a negative regulator of glucose homeostasis and is involved in energy metabolism, inflammation, ER-stress, mitochondrial dysfunction, and apoptotic pathways. GSK3B is a critical component of the Wnt/beta-catenin signaling pathway, and its activity results in the inhibition of Wnt signaling by mediating the degradation of beta-catenin. GSK3B is also involved in the Hedgehog, Notch, and insulin signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK3B
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal, pH 7.3
Gene GSK3B
Gene Accession No. P18266, P49841, Q9WV60
Gene Alias 7330414F15Rik, 8430431H08Rik, C86142, EC 2.7.11.26, FA, Factor A, fb68h05, fk80d11, glycogen synthase kinase 3, glycogen synthase kinase 3 beta, glycogen synthase kinase 3 beta L homeolog, glycogen synthase kinase 3 beta variant 1, glycogen synthase kinase 3 beta variant 2, glycogen synthase kinase 3 beta variant 3, glycogen synthase kinase 3 beta variant 4, glycogen synthase kinase 3 beta variant 5, glycogen synthase kinase 3 beta, genome duplicate a, glycogen synthase kinase-3 beta, glycogen synthase kinase-3 beta splice variant X1, glycogen synthase kinase-3 beta splice variant X2, glycogen synthase kinase-3 beta splice variant X3, GSK 3 beta, GSK 3B, gsk3, GSK-3, GSK3 beta, GSK-3 beta, GSK-3 beta GSK3B, GSK-3[b], Gsk3b, GSK-3b, gsk3b.L, gsk3ba, gsk3beta, GSK-3beta, gsk3-beta, GSK3beta isoform, intracellular kinase, pp37, serine/threonine-protein kinase GSK3B, wu:fb68h05, wu:fk80d11, XELAEV_180112401mg, xgsk-3, Xgsk-3 protein
Gene Symbols GSK3B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 350-420 of GSK3&beta, (NP_0011396281)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2932, 56637, 84027
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.