Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSPT2 Antibody (OTI2G9), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271768PEATT594
Description
GSPT2 Monoclonal specifically detects GSPT2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
GSPT2 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
GSPT2 | |
Full length recombinant protein of human GSPT2 (NP_060564) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Flow Cytometry | |
OTI2G9 | |
Flow Cytometry | |
eRF3b, ERF3BGST2, eukaryotic peptide chain release factor GTP-binding subunit ERF3B, Eukaryotic peptide chain release factor subunit 3b, FLJ10441, G1 to S phase transition 2, G1 to S phase transition protein 2 homolog | |
Mouse | |
Affinity Purified | |
Core ESC Like Genes, Stem Cell Markers | |
23708.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction