Learn More
Invitrogen™ GSPT2 Monoclonal Antibody (OTI2G9)

Description
GSPT2 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | GSPT2 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2G9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GSPT2 |
| Gene Accession No. | Q8IYD1 |
| Gene Alias | eRF3b; Eukaryotic peptide chain release factor GTP-binding subunit ERF3B; eukaryotic peptide chain release factor subunit 3b; FLJ10441; G1 to S phase transition 2; G1 to S phase transition protein 2 homolog; GSPT2; GST2; RGD1563213 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.