Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSPT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544324
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544324 Supplier Invitrogen™ Supplier No. PA544324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MALEESWEHS KEVSEAEPGG GSSGDSGPPE ESGQEMMEEK EEIRKSKSVI Sequence homology: Horse: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rat: 86%.

ERF3B encodes a GTPase that belongs to the GTP-binding elongation factor family. The encoded protein is a polypeptide release factor that complexes with eukaryotic peptide chain release factor 1 to mediate translation termination. This protein may also be involved in mRNA stability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSPT2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GSPT2
Gene Accession No. Q8IYD1
Gene Alias eRF3b; Eukaryotic peptide chain release factor GTP-binding subunit ERF3B; eukaryotic peptide chain release factor subunit 3b; FLJ10441; G1 to S phase transition 2; G1 to S phase transition protein 2 homolog; GSPT2; GST2; RGD1563213
Gene Symbols GSPT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GSPT2 (aa 144-193).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23708
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.