Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSR Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588332
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588332 Supplier Invitrogen™ Supplier No. PA588332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MISTNCTEEL ENAGVEVLKF SQVKEVKKTL SGLEVSMVTA VPGRLPVMTM IPDVDCLLWA IGRVPNTKDL SLNKLGIQTD DKGHIIVDEF QNTNVKGIYA VGDVCGKALL TPVAIAAGRK LAHRLFEYKE DSKLDYNNIP TVVFSHPPIG TVGLTEDEAI HKYGIENVKT YSTSFTPMYH AVTKRKTKCV MKMVCANKEE KVVGIHMQGL GCDEMLQGFA VAVKMGATKA DFDNTVAIHP TSSEELVTLR; Positive Samples: LO2, HeLa, A-549, Jurkat, Mouse brain, Mouse heart, Mouse kidney; Cellular Location: Cytoplasm, Mitochondrion.

This gene encodes a member of the class-I pyridine nucleotide-disulfide oxidoreductase family. This enzyme is a homodimeric flavoprotein. It is a central enzyme of cellular antioxidant defense, and reduces oxidized glutathione disulfide (GSSG) to the sulfhydryl form GSH, which is an important cellular antioxidant. Rare mutations in this gene result in hereditary glutathione reductase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSR
Applications ELISA, Western Blot, Western Blot
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Gsr
Gene Accession No. P00390, P47791
Gene Alias AI325518; D8Ertd238e; epididymis luminal protein 75; epididymis secretory sperm binding protein Li 122m; GLUR; glutathione reductase; glutathione reductase 1; glutathione reductase, mitochondrial; glutathione S-reductase; glutathione-disulfide reductase; GR; Gr1; Gr-1; GRase; GRD1; GSR; HEL-75; HEL-S-122m; hypothetical protein LOC553575; zgc:110010
Gene Symbols Gsr
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 273-522 of human GSR (NP_0006282).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14782, 2936
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.