Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSR Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
Antigen | GSR |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119144
|
Invitrogen™
PA5119144 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121944
|
Invitrogen™
PA5121944 |
20 μL |
Each for $178.40
|
|
|||||
Description
GSR Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the class-I pyridine nucleotide-disulfide oxidoreductase family. This enzyme is a homodimeric flavoprotein. It is a central enzyme of cellular antioxidant defense, and reduces oxidized glutathione disulfide (GSSG) to the sulfhydryl form GSH, which is an important cellular antioxidant. Rare mutations in this gene result in hereditary glutathione reductase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been found.Specifications
GSR | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
Gsr | |
AI325518; D8Ertd238e; epididymis luminal protein 75; epididymis secretory sperm binding protein Li 122m; GLUR; glutathione reductase; glutathione reductase 1; glutathione reductase, mitochondrial; glutathione S-reductase; glutathione-disulfide reductase; GR; Gr1; Gr-1; GRase; GRD1; GSR; HEL-75; HEL-S-122m; hypothetical protein LOC553575; zgc:110010 | |
Gsr | |
Primary | |
Antigen Affinity Chromatography |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P00390 | |
2936 | |
Recombinant protein Glutathione Reductase. The antigen corresponds to amino acid range 44-522 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title