Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562976
Description
Immunogen sequence: NLYGEEMVQA LKQLKDSEER ASYILMEKIE PEPFENCLLR PGSPARVVQC ISELGIFGVY VRQEKTLVMN KHVGHLLRTK AI Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.
Glutathione is important for a variety of biological functions, including protection of cells from oxidative damage by free radicals, detoxification of xenobiotics, and membrane transport. The protein encoded by this gene functions as a homodimer to catalyze the second step of glutathione biosynthesis, which is the ATP-dependent conversion of gamma-L-glutamyl-L-cysteine to glutathione. Defects in this gene are a cause of glutathione synthetase deficiency.
Specifications
GSS | |
Polyclonal | |
Unconjugated | |
GSS | |
AI314904; Glutathione synthase; glutathione synthetase; GS-A/GS-B; GSH S; GSH synthetase; GSHB; GSHS; GSH-S; GSS; HEL-S-64p; HEL-S-88n; MGC14098; OTTHUMP00000030711 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2937 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P48637 | |
GSS | |
Recombinant protein corresponding to Human GSS. Recombinant protein control fragment (Product #RP-99312). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction