Learn More
Cytiva GST Gene Fusion System pGEX Vectors
Catalog No. 45002275 Shop All Cytiva Products
Change view
Click to view available options
Kit Contents:
2T
4T-1
4T-3
6P-1
6P-2
6P-3
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Kit Contents
45-002-275 6P-1
45-002-271 4T-1
45-002-276 6P-2
45-002-277 6P-3
45-002-278 2T
45-002-282 4T-3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 45-002-275 Supplier Cytiva Supplier No. 28954648
Add to Cart
Add to Cart
Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schistosoma japonicum GST
  • Offers a tac promoter for chemically inducible, high-level expression of GST-tagged recombinant proteins
  • Offers an internal laq1q gene for use in any E. coli host
  • With PreScission™ Protease, Thrombin, or Factor Xa recognition sites for cleaving the target protein from the fusion product
  • The vectors provide all three translational reading frames beginning with the EcoRI restriction site
  • Can use a wide variety of E. coli host strains cloning and expression
  • pGEX-1lT EcoRI/BAP is an EcoR1 pre cut vector, phosphatase treated to prevent self ligation, ready for gene insertion
  • pGEX-2TK will introduce a specific kinase site allowing the fusion protein to be labelled with [γ-32P]ATP using protein kinase
TRUSTED_SUSTAINABILITY
Specifications
Kit Contents 6P-1
Quantity 25 μg
For Use With (Application) Designed for inducible, high-level intracellular expression of genes or gene fragments as fusions with Schisto soma japonicum GST
Type pGEX Vector