Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Monoclonal Antibody (2F10B9), THE™, GenScript™

Catalog No. 50272995
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
200 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-272-995 100 μg
50-272-997 200 μg
50-272-998 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-272-995 Supplier Genscript Corporation Supplier No. A00865100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute the lyophilized powder with deionized water (or equivalent) to an antibody concentration of 0.5 mg/mL. Schistosoma japonicum glutathione S-transferase (GST) is a 26,000 Da enzyme that conjugates reduced glutathione to hydrophobic electrophiles. GST is an ideal fusion partner for production of foreign proteins in commercially available expression systems because it can be expressed in high levels in E. coli. The fusion proteins can be purified to homogenity in a single step as the GST portion of the protein binds tightly to immobilized glutathione. Elution can be accomplished using free glutathione. The GST portion of the fusion protein can be separated from the protein of interest by using site-specific proteases. Antibodies directed against GST can be helpful in detecting the fusion protein during purification and to detect cleavage of GST from the protein of interest.

GST Monoclonal antibody specifically detects GST in Tag samples. It is validated for ELISA, Immunoprecipitation, Western Blot

Specifications

Antigen GST
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2F10B9
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide; pH 7.4
Gene Alias activator protein 1; AP1; AP-1; cancer/testis antigen 48; c-Jun; CT48; deafness, X-linked 7; DFN7; enhancer-binding protein AP1; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase M1; glutathione S-transferase mu 1; Glutathione S-transferase P; glutathione S-transferase pi 1; Glutathione-S-Transferase; gonad-specific transporter; GST; GST class-mu 1; GST class-pi; GST HB subunit 4; GST protein; GST Tag; GST1; GST3; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP; GSTP1; GSTP1-1; GTH4; GTM1; H-B; HB subunit 4; HEL-S-22; JUN; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; MU; MU-1; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; p39; PI; Proto-oncogene c-Jun; S-(hydroxyalkyl)glutathione lyase; SLC21A19
Host Species Mouse
Immunogen Recombinant GST protein (26 kD)
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Lyophilized
Isotype IgG1 κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.