Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Mouse anti-S. japonicum, ATTO 488, Clone: 3E2, Abnova™

Catalog No. 89339077 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-077 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-077 Supplier Abnova Corporation Supplier No. MAB19792
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

GST Monoclonal antibody specifically detects GST in S. japonicum samples. It is validated for WB, ELISA.

Specifications

Antigen GST
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3E2
Conjugate Atto 488
Host Species Mouse
Immunogen Glutathione-S-Transferase (GST) tagged protein
Quantity 100 μg
Primary or Secondary Primary
Target Species S. japonicum
Content And Storage Store at -20°C
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.