Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GST Omega 1 140A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531185
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531185 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531185 Supplier Invitrogen™ Supplier No. PA531185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HepG2, mouse colon, rat liver. Predicted reactivity: Pig (82%), Rhesus Monkey (97%), Chimpanzee (98%), Bovine (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is an omega class glutathione S-transferase (GST) with glutathione-dependent thiol transferase and dehydroascorbate reductase activities. GSTs are involved in the metabolism of xenobiotics and carcinogens. The encoded protein acts as a homodimer and is found in the cytoplasm. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GST Omega 1 140A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Gsto1
Gene Accession No. O09131, P78417, Q9Z339
Gene Alias AA407097; AI194287; AU018802; epididymis secretory protein Li 21; glutathione S-transferase like; glutathione S-transferase omega 1; glutathione S-transferase omega 1-1; Glutathione S-transferase omega-1; Glutathione transferase omega 1 (GSTO 1-1) (p28); glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase like; GSTO 1-1; Gsto1; GSTO-1; GSTTLP28; GSTX; Gtsttl; HEL-S-21; MMA(V) reductase; Monomethylarsonic acid reductase; P28; RP11-99N20.1; S-(Phenacyl)glutathione reductase; SPG-R
Gene Symbols Gsto1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GSTO1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114846, 14873, 9446
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.