Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GST Omega 1 140A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583382
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583382 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583382 Supplier Invitrogen™ Supplier No. PA583382
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WFERLEAMKL NECVDHTPKL KLWMAAMKED PTVSALLTSE KDWQGFLELY LQNSPEACDY GL Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 66%.

The protein encoded by this gene is an omega class glutathione S-transferase (GST) with glutathione-dependent thiol transferase and dehydroascorbate reductase activities. GSTs are involved in the metabolism of xenobiotics and carcinogens. The encoded protein acts as a homodimer and is found in the cytoplasm. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GST Omega 1 140A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gsto1
Gene Accession No. P78417
Gene Alias AA407097; AI194287; AU018802; epididymis secretory protein Li 21; glutathione S-transferase like; glutathione S-transferase omega 1; glutathione S-transferase omega 1-1; Glutathione S-transferase omega-1; Glutathione transferase omega 1 (GSTO 1-1) (p28); glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase like; GSTO 1-1; Gsto1; GSTO-1; GSTTLP28; GSTX; Gtsttl; HEL-S-21; MMA(V) reductase; Monomethylarsonic acid reductase; P28; RP11-99N20.1; S-(Phenacyl)glutathione reductase; SPG-R
Gene Symbols Gsto1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GSTO1. Recombinant protein control fragment (Product #RP-97118).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9446
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.