Learn More
Description
Specifications
Specifications
| Antigen | GST Tag |
| Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Formulation | Stabilized solution in 50% glycerol |
| Host Species | Mouse |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Test Specificity | 220aa GST Protein, precise epitope not determined |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.