Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GST Tag Monoclonal Antibody (vpg66), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA181040
Description
Mouse anti GST antibody, clone vpg66 reacts with Glutathione-S-transferase.
GST (Glutathione S-transferase) is a 26kDa protein, present in eukaryotes and in prokaryotes, where they catalyze a variety of reactions. Glutathione Stransferase is used to create the GST gene fusion system. This GST-fusion protein can then be purified from cells via its high affinity for glutathione. Fusion Proteins offer an important biological assay for direct protein-to-protein interactions. The GST gene fusion system has been introduced into numerous expression vectors.
Specifications
GST Tag | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.09% sodium azide | |
Mouse | |
purified | |
RUO | |
Tag | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
vpg66 | |
Biotin | |
activator protein 1; AP1; AP-1; cancer/testis antigen 48; c-Jun; CT48; deafness, X-linked 7; DFN7; enhancer-binding protein AP1; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase M1; glutathione S-transferase mu 1; Glutathione S-transferase P; glutathione S-transferase pi 1; Glutathione-S-Transferase; gonad-specific transporter; GST; GST class-mu 1; GST class-pi; GST HB subunit 4; GST protein; GST Tag; GST1; GST3; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP; GSTP1; GSTP1-1; GTH4; GTM1; H-B; HB subunit 4; HEL-S-22; JUN; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; MU; MU-1; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; p39; PI; Proto-oncogene c-Jun; S-(hydroxyalkyl)glutathione lyase; SLC21A19; SLCO6A1; Solute carrier family 21 member 19; solute carrier organic anion transporter family member 6A1; solute carrier organic anion transporter family, member 6A1; testicular tissue protein Li 174; testis-specific organic anion transporter; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog | |
GST from Schistosoma japonicum. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction