Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST tag Mouse, Clone: IL382.43, Abnova™

Catalog No. 89035471 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-471 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-471 Supplier Abnova Corporation Supplier No. MAB14067

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against recombinant GST tag.

  • High specificity
  • High reactivity

Specifications

Antigen GST tag
Applications ELISA, Western Blot
Classification Monoclonal
Clone IL382.43
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant GST tag.
Dilution ELISA Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide).
Host Species Mouse
Immunogen Recombinant protein corresponding to GST tag.
Purification Method Protein G affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.