Learn More
GST-tag Rabbit, Polyclonal, GenScript™
Rabbit Polyclonal Antibody
Supplier: Genscript Corporation A0013040

Description
Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 0.5 mg/mL. Rabbit Anti-GST-tag [HRP]; GST-tag [HRP] Antibody
GST-tag Polyclonal antibody specifically detects GST-tag in Tag samples. It is validated for ELISA, Western BlotSpecifications
GST-tag | |
Polyclonal | |
PBS with 1% BSA and 0.01% thimerosal | |
Rabbit | |
Affinity chromatography | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Tag |
ELISA, Western Blot | |
Unconjugated | |
activator protein 1; AP1; AP-1; cancer/testis antigen 48; c-Jun; CT48; deafness, X-linked 7; DFN7; enhancer-binding protein AP1; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase M1; glutathione S-transferase mu 1; Glutathione S-transferase P; glutathione S-transferase pi 1; Glutathione-S-Transferase; gonad-specific transporter; GST; GST class-mu 1; GST class-pi; GST HB subunit 4; GST protein; GST Tag; GST1; GST3; GSTM1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; GSTP; GSTP1; GSTP1-1; GTH4; GTM1; H-B; HB subunit 4; HEL-S-22; JUN; Jun activation domain binding protein; jun oncogene; jun proto-oncogene; Jun proto-oncogene, AP-1 transcription factor subunit; MU; MU-1; OATP6A1; OATP-I; OATPY; Organic anion-transporting polypeptide 6A1; Organic anion-transporting polypeptide I; p39; PI; Proto-oncogene c-Jun; S-(hydroxyalkyl)glutathione lyase; SLC21A19; SLCO6A1; Solute carrier family 21 member 19; solute carrier organic anion transporter family member 6A1; solute carrier organic anion transporter family, member 6A1; testicular tissue protein Li 174; testis-specific organic anion transporter; transcription factor AP-1; V-jun avian sarcoma virus 17 oncogene homolog; v-jun sarcoma virus 17 oncogene homolog | |
Recombinant Glutathione S-Transferase (GST) from Schistosoma japonicum | |
40 μg | |
Primary | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.