Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTA1 Monoclonal Antibody (2-2#), Invitrogen™
GREENER_CHOICE

Catalog No. PIMA543788
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543788 100 μL
PIMA543789 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543788 Supplier Invitrogen™ Supplier No. MA543788
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

This gene encodes a member of a family of enzymes that function to add glutathione to target electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins, and products of oxidative stress. This action is an important step in detoxification of these compounds. This subfamily of enzymes has a particular role in protecting cells from reactive oxygen species and the products of peroxidation. Polymorphisms in this gene influence the ability of individuals to metabolize different drugs. This gene is located in a cluster of similar genes and pseudogenes on chromosome 6. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTA1
Applications Western Blot
Classification Monoclonal
Clone 4#
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GSTA1
Gene Accession No. P08263
Gene Alias 13-hydroperoxyoctadecadienoate peroxidase; Androst-5-ene-3,17-dione isomerase; class alpha; glutathione S-alkyltransferase A1; glutathione S-aryltransferase A1; glutathione S-transferase; glutathione S-transferase 2; glutathione S-transferase A1; Glutathione S-transferase A1, N-terminally processed; glutathione S-transferase A3 subunit; glutathione S-transferase A5; glutathione S-transferase alpha 1; glutathione S-transferase alpha M14; Glutathione S-transferase alpha M14, N-terminally processed; Glutathione S-transferase alpha-3; glutathione S-transferase Ha subunit 1; glutathione S-transferase Ya; Glutathione S-transferase Ya1; glutathione S-transferase Yc-1; glutathione S-transferase Yc1 subunit; glutathione S-transferase, alpha 1 (Ya); Glutathione-S-transferase alpha type (Ya); GST; GST 2-2; GST A3-3; GST AA; GST class-alpha; GST class-alpha member 1; GST HA subunit 1; GST Yc1; GST2; Gst2-1; Gsta; GSTA1; GSTA1-1; Gsta3; Gsta5; GST-epsilon; Gstya; Gstyc1; GTH1; OTTMUSG00000031890; RP11-392A23.1; S-(hydroxyalkyl)glutathione lyase A1; testicular tissue protein Li 80; Yc1
Gene Symbols GSTA1
Host Species Mouse
Immunogen Ala2-Phe222 (Accession P08263), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2938
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.