Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100255 Supplier Invitrogen™ Supplier No. PA5100255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GSTA2.

Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. These enzymes function in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding these enzymes are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of some drugs. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-tranferase belonging to the alpha class. The alpha class genes, located in a cluster mapped to chromosome 6, are the most abundantly expressed glutathione S-transferases in liver. In addition to metabolizing bilirubin and certain anti-cancer drugs in the liver, the alpha class of these enzymes exhibit glutathione peroxidase activity thereby protecting the cells from reactive oxygen species and the products of peroxidation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTA2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gsta2
Gene Accession No. P04903, P09210, P10648
Gene Alias 13-hydroperoxyoctadecadienoate peroxidase; Androst-5-ene-3,17-dione isomerase; glutathione S-alkyltransferase A2; glutathione S-aralkyltransferase A2; glutathione S-aryltransferase A2; glutathione S-transferase A2; glutathione S-transferase alpha 2; glutathione S-transferase alpha-1; Glutathione S-transferase GT41A; glutathione S-transferase Ya subunit; glutathione S-transferase Ya-1; glutathione S-transferase, alpha 2 (Yc2); Glutathione-S-transferase alpha type (Yc2); glutathione-S-transferase, alpha type2; GST 1-1; GST 1a-1a; GST A1-1; GST B; GST class-alpha member 2; GST HA subunit 2; GST Ya1; GST2; Gst2-2; Gsta1; Gsta2; GSTA2-2; Gstc2; Gstc-2; GST-gamma; GTA2; GTH2; Ligandin; liver GST2; S-(hydroxyalkyl)glutathione lyase A2; testis tissue sperm-binding protein Li 59n
Gene Symbols Gsta2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GSTA2(Accession P09210), corresponding to amino acid residues E97-Y147.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14858, 24422, 2939
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.