Learn More
Invitrogen™ GSTA2 Polyclonal Antibody

Description
The antibody detects endogenous levels of total GSTA2 protein.
Specifications
Specifications
| Antigen | GSTA2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Gsta2 |
| Gene Accession No. | P09210 |
| Gene Alias | 13-hydroperoxyoctadecadienoate peroxidase; Androst-5-ene-3,17-dione isomerase; glutathione S-alkyltransferase A2; glutathione S-aralkyltransferase A2; glutathione S-aryltransferase A2; glutathione S-transferase A2; glutathione S-transferase alpha 2; glutathione S-transferase alpha-1; Glutathione S-transferase GT41A; glutathione S-transferase Ya subunit; glutathione S-transferase Ya-1; glutathione S-transferase, alpha 2 (Yc2); Glutathione-S-transferase alpha type (Yc2); glutathione-S-transferase, alpha type2; GST 1-1; GST 1a-1a; GST A1-1; GST B; GST class-alpha member 2; GST HA subunit 2; GST Ya1; GST2; Gst2-2; Gsta1; Gsta2; GSTA2-2; Gstc2; Gstc-2; GST-gamma; GTA2; GTH2; Ligandin; liver GST2; S-(hydroxyalkyl)glutathione lyase A2; testis tissue sperm-binding protein Li 59n |
| Gene Symbols | Gsta2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.