Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304544100UL
Description
GSTA2 Polyclonal specifically detects GSTA2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GSTA2 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 2939 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| EC 2.5.1.18, glutathione S-aralkyltransferase A2, glutathione S-aryltransferase A2, glutathione S-transferase 2, glutathione S-transferase A2, glutathione S-transferase alpha 2, GST class-alpha member 2, GST HA subunit 2, GST, class alpha, 2, GST2glutathione S-alkyltransferase A2, GSTA2-2, GST-gamma, GTA2, GTH2, HA subunit 2, liver GST2, MGC10525, S-(hydroxyalkyl)glutathione lyase A2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-222 of human GSTA2 (NP_000837.3). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction