Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119147
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119147 200 μL
PIPA5121942 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119147 Supplier Invitrogen™ Supplier No. PA5119147
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GSTA3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. These enzymes are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-tranferase belonging to the alpha class genes that are located in a cluster mapped to chromosome 6. Genes of the alpha class are highly related and encode enzymes with glutathione peroxidase activity. However, during evolution, this alpha class gene diverged accumulating mutations in the active site that resulted in differences in substrate specificity and catalytic activity. The enzyme encoded by this gene catalyzes the double bond isomerization of precursors for progesterone and testosterone during the biosynthesis of steroid hormones. An additional transcript variant has been identified, but its full length sequence has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTA3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Gsta3
Gene Accession No. P30115
Gene Alias glutathione S-alkyltransferase A3; glutathione S-aralkyltransferase A3; glutathione S-aryltransferase A3; glutathione S-transferase A3; glutathione S-transferase A3 subunit; glutathione S-transferase A3-3; glutathione S-transferase A5; glutathione S-transferase alpha 1; glutathione S-transferase alpha 3; glutathione S-transferase alpha-3; Glutathione S-transferase alpha-5; glutathione S-transferase Ya3; Glutathione S-transferase Yc; glutathione S-transferase Yc-1; glutathione S-transferase Yc1 subunit; Glutathione S-transferase Yc-2; glutathione S-transferase Yc2 subunit; glutathione S-transferase, alpha 3; Glutathione-S-transferase alpha type (Ya); GST 2-2; GST A3-3; GST A5-5; GST AA; GST class-alpha member 3; GST Yc1; GST Yc2; Gst2-3; Gsta1; GSTA3; GSTA3-3; Gsta5; Gstyc; Gstyc1; Gstyc2; GTA3; MGC22232; S-(hydroxyalkyl)glutathione lyase A3; Yc1; Yc2
Gene Symbols Gsta3
Host Species Rabbit
Immunogen Recombinant protein Glutathione S Transferase Alpha 3. The antigen corresponds to amino acid range 1-221 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14859
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.