Learn More
Invitrogen™ GSTA3 Polyclonal Antibody
Description
GSTA3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | GSTA3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | Gsta3 |
| Gene Accession No. | P30115 |
| Gene Alias | glutathione S-alkyltransferase A3; glutathione S-aralkyltransferase A3; glutathione S-aryltransferase A3; glutathione S-transferase A3; glutathione S-transferase A3 subunit; glutathione S-transferase A3-3; glutathione S-transferase A5; glutathione S-transferase alpha 1; glutathione S-transferase alpha 3; glutathione S-transferase alpha-3; Glutathione S-transferase alpha-5; glutathione S-transferase Ya3; Glutathione S-transferase Yc; glutathione S-transferase Yc-1; glutathione S-transferase Yc1 subunit; Glutathione S-transferase Yc-2; glutathione S-transferase Yc2 subunit; glutathione S-transferase, alpha 3; Glutathione-S-transferase alpha type (Ya); GST 2-2; GST A3-3; GST A5-5; GST AA; GST class-alpha member 3; GST Yc1; GST Yc2; Gst2-3; Gsta1; GSTA3; GSTA3-3; Gsta5; Gstyc; Gstyc1; Gstyc2; GTA3; MGC22232; S-(hydroxyalkyl)glutathione lyase A3; Yc1; Yc2 |
| Gene Symbols | Gsta3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.