Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTA4 Antibody (OTI3F5), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270856F
Description
GSTA4 Monoclonal specifically detects GSTA4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
GSTA4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
GSTA4 | |
Full length recombinant protein of human GSTA4 (NP_001503) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry, Immunohistochemistry | |
OTI3F5 | |
Flow Cytometry, Immunohistochemistry | |
DKFZp686D21185, EC 2.5.1.18, glutathione S-alkyltransferase A4, glutathione S-aralkyltransferase A4, glutathione S-aryltransferase A4, glutathione S-transferase A4, Glutathione S-transferase A4-4, glutathione S-transferase alpha 4, glutathione transferase A4-4, GST class-alpha member 4, GSTA4-4, GTA4, S-(hydroxyalkyl)glutathione lyase A4 | |
Mouse | |
Affinity Purified | |
metabolism | |
2941.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction