Learn More
Invitrogen™ GSTA4 Monoclonal Antibody (OTI3F5)

Description
GSTA4 Monoclonal Antibody for Western Blot, IHC (P), Flow
Specifications
Specifications
| Antigen | GSTA4 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3F5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GSTA4 |
| Gene Accession No. | O15217 |
| Gene Alias | glutathione S-alkyltransferase A4; glutathione S-aralkyltransferase A4; glutathione S-aryltransferase A4; glutathione S-transferase 5.7; Glutathione S-transferase A4; glutathione S-transferase A4-4; glutathione S-transferase alpha 4; glutathione S-transferase, alpha 4; glutathione transferase A4-4; GST 5.7; GST A4-4; GST class-alpha member 4; GST5.7; Gsta; GSTA4; GSTA4-4; GTA4; mGsta4; S-(hydroxyalkyl)glutathione lyase A4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.