Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119145
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119145 200 μL
PIPA5121941 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119145 Supplier Invitrogen™ Supplier No. PA5119145
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GSTA4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. This isozyme has a high catalytic efficiency with 4-hydroxyalkenals such as 4-hydroxynonenal (4-HNE).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTA4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GSTA4
Gene Accession No. P14942
Gene Alias glutathione S-alkyltransferase A4; glutathione S-aralkyltransferase A4; glutathione S-aryltransferase A4; glutathione S-transferase 5.7; Glutathione S-transferase A4; glutathione S-transferase A4-4; glutathione S-transferase alpha 4; glutathione S-transferase, alpha 4; glutathione transferase A4-4; GST 5.7; GST A4-4; GST class-alpha member 4; GST5.7; Gsta; GSTA4; GSTA4-4; GTA4; mGsta4; S-(hydroxyalkyl)glutathione lyase A4
Gene Symbols GSTA4
Host Species Rabbit
Immunogen Recombinant protein Glutathione S Transferase A4. The antigen corresponds to amino acid range 2-222 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 300850
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.