Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119148
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119148 200 μL
PIPA5121940 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119148 Supplier Invitrogen™ Supplier No. PA5119148
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GSTA5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

The glutathione S-transferases catalyze the conjugation of reduced glutathiones and a variety of electrophiles, including many known carcinogens and mutagens. The cytosolic GSTs belong to a large superfamily, with members located on different chromosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTA5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene GSTA5
Gene Accession No. P46418, Q7RTV2
Gene Alias glutathione S-transferase A5; Glutathione S-transferase A5-5; glutathione S-transferase alpha 5; glutathione transferase A5; GST class-alpha member 5; GSTA5
Gene Symbols GSTA5
Host Species Rabbit
Immunogen Recombinant protein Glutathione S Transferase Alpha 5. The antigen corresponds to amino acid range 1-221 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221357, 494499
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.