Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTCD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546016 Supplier Invitrogen™ Supplier No. PA546016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LRESSDQPPT IPVEILQLEK KLSEPVRVHN DDKLRRQKLK QQKADGVGPP Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 85%; Horse: 86%; Human: 100%; Mouse: 92%; Pig: 86%; Rabbit: 100%; Rat: 100%.

GSTCD (glutathione S-transferase, C-terminal domain containing) is a 633 amino acid protein belonging to the GSTCD family and contains one GST C-terminal domain. The gene encoding GSTCD maps to human chromosome 4, which represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTCD
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Gstcd
Gene Accession No. Q8NEC7
Gene Alias 4933434L15Rik; glutathione S-transferase C-terminal domain containing; glutathione S-transferase C-terminal domain-containing protein; glutathione S-transferase, C-terminal domain containing; GSTCD; RGD1311822
Gene Symbols Gstcd
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GSTCD (aa 162-211).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79807
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.