Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSTCD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111535
Description
Immunogen sequence: SEKLVEFPLL ASWYQRIQEV PGVKTAASKC GIQFLHLPKL LTTSTEQHPN LCEVPGVEEQ SDPLFIGGPR PTMAKLMEKG IEVMFSPHPC PTWTLDW.
GSTCD (glutathione S-transferase, C-terminal domain containing) is a 633 amino acid protein belonging to the GSTCD family and contains one GST C-terminal domain. The gene encoding GSTCD maps to human chromosome 4, which represents approximately 6% of the human genome and contains nearly 900 genes. Notably, the Huntingtin gene, which is found to encode an expanded glutamine tract in cases of Huntington's disease, is on chromosome 4. FGFR-3 is also encoded on chromosome 4 and has been associated with thanatophoric dwarfism, achondroplasia, Muenke syndrome and bladder cancer.
Specifications
GSTCD | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NEC7 | |
Gstcd | |
Recombinant Protein corresonding to Human GSTCD. Recombinant protein control fragment (Product #RP-107094). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry | |
Unconjugated | |
Gstcd | |
4933434L15Rik; glutathione S-transferase C-terminal domain containing; glutathione S-transferase C-terminal domain-containing protein; glutathione S-transferase, C-terminal domain containing; GSTCD; RGD1311822 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79807 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction