Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535381 Supplier Invitrogen™ Supplier No. PA535381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody has been tested in 3T3-L1 adipocytes and mouse white and brown adipose tissue lysate samples. It contains enough material to conduct 10 mini-Western blots. It is recommended to add 1% BSA to the antibody dilution buffer.

GSTK1 encodes a member of the kappa class of the glutathione transferase superfamily of enzymes that function in cellular detoxification. The encoded protein is localized to the peroxisome and catalyzes the conjugation of glutathione to a wide range of hydrophobic substates facilitating the removal of these compounds from cells. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTK1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 40% glycerol and no preservative
Gene GSTK1
Gene Accession No. Q9DCM2, Q9Y2Q3
Gene Alias 0610025I19Rik; AW260476; DsbA-L; glutathione S-transferase class kappa; glutathione S-transferase k1; glutathione S-transferase kappa 1; Glutathione S-transferase subunit 13; glutathione S-transferase subunit 13 homolog; glutathione S-transferase, mitochondrial; GST; GST 13-13; GST class-kappa; GST13; GST13-13; Gstk1; GSTK1-1; GSTkappa; HDCMD47P; hGSTK1; mGSTK1; rGSTK1
Gene Symbols GSTK1
Host Species Rabbit
Immunogen Full length recombinant mouse DsbA-L protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 373156, 76263
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.