Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTM1/GSTM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518335
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518335 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518335 Supplier Invitrogen™ Supplier No. PA518335
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.

Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13.3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Null mutations of this class mu gene have been linked with an increase in a number of cancers, likely due to an increased susceptibility to environmental toxins and carcinogens. Multiple protein isoforms are encoded by transcript variants of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTM1/GSTM2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene GSTM1
Gene Accession No. P09488, P28161
Gene Alias glutathione S-alkyltransferase; glutathione S-alkyltransferase M2; glutathione S-aralkyltransferase; glutathione S-aralkyltransferase M2; glutathione S-aryltransferase; glutathione S-aryltransferase M2; glutathione S-transferase 4; glutathione S-transferase GT8.7; glutathione S-transferase M1; glutathione S-transferase M2; glutathione S-transferase M2 (muscle); glutathione S-transferase Mu 1; Glutathione S-transferase Mu 2; glutathione S-transferase mu 2 (muscle); glutathione S-transferase pmGT2; Glutathione S-transferase Yb-1; glutathione S-transferase Yb-1 subunit; glutathione S-transferase Yb-2; glutathione S-transferase Yb2 subunit; glutathione S-transferase, mu 1; glutathione S-transferase, mu 2; glutathione-S-transferase mu type 1 (Yb1); Glutathione-S-transferase mu type 2 (Yb2); glutathione-S-transferase, mu 1; glutathione-S-transferase, mu 2; glutathione-S-transferase, mu type 1 (Yb1); Glutathione-S-transferase, mu type 2 (Yb2); GST 1-1; GST 3-3; GST 4-4; GST 5-5; GST class-mu 1; GST class-mu 2; GST HB subunit 4; GST Yb1; GST Yb2; GST, muscle; GST1; GST4; GSTA3; GSTA4; Gstb1; Gstb-1; Gstb2; Gstb-2; GSTM; Gstm1; GSTM1-1; GSTM1a-1a; GSTM1b-1b; Gstm2; GSTM2-2; GTH4; GTHMUS; GTM1; H-B; HB subunit 4; MGC117303; MGC26563; MU; MU-1; pmGT10; S-(hydroxyalkyl)glutathione lyase; S-(hydroxyalkyl)glutathione lyase M2
Gene Symbols GSTM1, GSTM2
Host Species Goat
Immunogen Synthetic peptide sequence (CVFSKMAVWGNK) corresponding to the C-terminus amino acids of GSTM1GSTM2 (aa 208-218).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2944, 2946
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.