Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTM2 Antibody (CPTC-GSTMu2-2), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308255MFV610
Description
GSTM2 Monoclonal antibody specifically detects GSTM2 in Human samples. It is validated for Flow CytometrySpecifications
GSTM2 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
CPTC-GSTMu2-2 | |
Flow Cytometry | |
glutathione S-transferase mu 2 (muscle), MGC117303, muscle, S-(hydroxyalkyl)glutathione lyase M2 | |
Recombinant human full-length GSTM2 protein (Uniprot: P28161) | |
0.1 mL | |
Endocrinology, Signal Transduction | |
2946 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction