Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTM2 Recombinant Rabbit Monoclonal Antibody (011)
GREENER_CHOICE

Catalog No. PIMA530476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530476 Supplier Invitrogen™ Supplier No. MA530476
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human GSTM2.

At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. GSTM2 is a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione.Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13.3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTM2
Applications ELISA
Classification Recombinant Monoclonal
Clone 011
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GSTM2
Gene Accession No. P28161
Gene Alias glutathione S-alkyltransferase M2; glutathione S-aralkyltransferase M2; glutathione S-aryltransferase M2; glutathione S-transferase 4; glutathione S-transferase M1; glutathione S-transferase M2; glutathione S-transferase M2 (muscle); glutathione S-transferase Mu 2; glutathione S-transferase mu 2 (muscle); glutathione S-transferase pmGT2; Glutathione S-transferase Yb-2; glutathione S-transferase Yb2 subunit; glutathione S-transferase, mu 2; Glutathione-S-transferase mu type 2 (Yb2); glutathione-S-transferase, mu 2; Glutathione-S-transferase, mu type 2 (Yb2); GST 4-4; GST 5-5; GST class-mu 2; GST Yb2; GST, muscle; GST4; GSTA4; Gstb2; Gstb-2; GSTM; GSTM2; GSTM2-2; GTHMUS; MGC117303; S-(hydroxyalkyl)glutathione lyase M2
Gene Symbols GSTM2
Host Species Rabbit
Immunogen Recombinant Human GSTM2 protein (Met1-Lys218).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2946
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.