Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTM3 Antibody (20C11.H5.E11), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP244175JF549
Description
GSTM3 Monoclonal antibody specifically detects GSTM3 in Human, Mouse samples. It is validated for ELISA, ImmunohistochemistrySpecifications
GSTM3 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
2947 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA, Immunohistochemistry | |
20C11.H5.E11 | |
ELISA, Immunohistochemistry | |
glutathione S-transferase mu 3 (brain), GST5Mu-3, MGC3310, MGC3704, S-(hydroxyalkyl)glutathione lyase M3 | |
This Protein A purified GSTM3 Antibody (20C11.H5.E11) was prepared by repeated immunizations in mice with GSTGSTM3 Protein (Uniprot: P21266) | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction