Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTM3 Antibody (CPTC-GSTMu3-1), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308256C
Description
GSTM3 Monoclonal antibody specifically detects GSTM3 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
GSTM3 | |
Monoclonal | |
DyLight 650 | |
glutathione S-transferase mu 3 (brain), GST5Mu-3, MGC3310, MGC3704, S-(hydroxyalkyl)glutathione lyase M3 | |
Recombinant full-length human GSTM3 protein (Uniprot: P21266) | |
100 μg | |
Cancer, Endocrinology, Signal Transduction | |
2947 | |
Store at 4°C in the dark. | |
IgG2a κ |
Immunohistochemistry (Paraffin), Protein Array | |
CPTC-GSTMu3-1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction