Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTM3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579338
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579338 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579338 Supplier Invitrogen™ Supplier No. PA579338
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, rat brain tissue, mouse testis tissue, mouse brain tissue, human Hela cell, human MCF-7 cell. IHC: human rectal cancer tissue, human lung cancer tissue, human testis tissue, mouse brain tissue, mouse testis tissue, rat brain tissue, rat testis tissue.

Glutathione S-transferase M3 (brain), also known as GSTM3, is an enzyme which in humans is encoded by theGSTM3 gene. Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13. 3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Mutations of this class mu gene have been linked with a slight increase in a number of cancers, likely due to exposure with environmental toxins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTM3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene GSTM3
Gene Accession No. P19639, P21266
Gene Alias brain GST; brain type mu-glutathione S-transferase; Fsc2; glutathione S-alkyltransferase M3; glutathione S-aralkyltransferase M3; glutathione S-aryltransferase M3; glutathione S-transferase GT9.3; glutathione S-transferase M3 (brain); glutathione S-transferase Mu 3; glutathione S-transferase mu 3 (brain); glutathione S-transferase, mu 3; glutathione S-transferase, Mu-3; GST class-mu 3; GST5; GSTB; GSTM3; GSTM3-3; GTM3; hGSTM3-3; mGSTM5; S-(hydroxyalkyl)glutathione lyase M3
Gene Symbols GSTM3
Host Species Rabbit
Immunogen E. coli-derived human GSTM3 recombinant protein (Position: E93-Q206).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14864, 2947
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.