Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTM4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP275088
Description
GSTM4 Polyclonal specifically detects GSTM4 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
GSTM4 | |
Polyclonal | |
Unconjugated | |
TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide | |
GSTM4 | |
Peptide with sequence DVSNQLARVCYSPD, from the internal region of the protein sequence according to NP_000841.1; NP_671489.1. | |
Antigen Affinity-purified | |
Primary | |
Human, Rat | |
Purified |
Western Blot, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/ml | |
Western Blot 1:100 - 1:2000, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:128000 | |
EC 2.5.1.18, glutathione S-alkyltransferase M4, glutathione S-aralkyltransferase M4, glutathione S-aryltransferase M4, glutathione S-transferase M4, glutathione S-transferase mu 4, GST class-mu 4, GSTM4-4, GST-Mu2, GTM4, GTS-Mu2, MGC131945, MGC9247, S-(hydroxyalkyl)glutathione lyase M4 | |
Goat | |
25.6 kDa | |
100 μg | |
2948.0 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction