Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100149 Supplier Invitrogen™ Supplier No. PA5100149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GSTM4.

Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13.3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Diversification of these genes has occurred in regions encoding substrate-binding domains, as well as in tissue expression patterns, to accommodate an increasing number of foreign compounds. Multiple transcript variants, each encoding a distinct protein isoform, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTM4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GSTM4
Gene Accession No. Q03013
Gene Alias 1110004G14Rik; glutathione S-alkyltransferase M4; glutathione S-aralkyltransferase M4; glutathione S-aryltransferase M4; glutathione S-transferase M4; glutathione S-transferase mu 3; glutathione S-transferase Mu 4; glutathione S-transferase, mu 4; glutathione-S-transferase, mu 4; GST class-mu 4; Gstb4; Gstb-4; GSTM4; GSTM4-4; GSTM7-7; GST-Mu2; GTM4; GTS-Mu2; S-(hydroxyalkyl)glutathione lyase M4; testis tissue sperm-binding protein Li 60n
Gene Symbols GSTM4
Host Species Rabbit
Immunogen A synthesized peptide derived from human GSTM4(Accession Q03013), corresponding to amino acid residues R82-P132.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14865, 2948
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.