Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GSTM5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527981
Description
Recommended positive controls: mouse brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13.3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Diversification of these genes has occurred in regions encoding substrate-binding domains, as well as in tissue expression patterns, to accommodate an increasing number of foreign compounds.
Specifications
GSTM5 | |
Polyclonal | |
Unconjugated | |
GSTM5 | |
fibrous sheath component 2; Fsc2; glutathione S-alkyltransferase M5; glutathione S-aralkyltransferase M5; glutathione S-aryltransferase M5; glutathione S-transferase M5; glutathione S-transferase Mu 5; glutathione S-transferase, mu 5; glutathione-S-transferase, mu 5; GST class-mu 5; Gstm3; GSTM5; GSTM5-5; GTM5; S-(hydroxyalkyl)glutathione lyase M5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14866, 2949 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P46439, P48774 | |
GSTM5 | |
Recombinant protein encompassing a sequence within the center region of human GSTM5. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction