Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116635 Supplier Invitrogen™ Supplier No. PA5116635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse heart, Mouse kidney, Mouse liver, Rat testis, Rat brain Immunogen sequence: LENQVMDNHM ELVRLCYDPD FEKLKPKYLE ELPEKLKLYS EFLGKRPWFA GDKITFVDFL AYDVLDMKRI FEPKCLDAFL NLKDFISRFE GLKKISAYMK SSQFLRGLLF GKSATWNSK.

Cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. This gene encodes a glutathione S-transferase that belongs to the mu class. The mu class of enzymes functions in the detoxification of electrophilic compounds, including carcinogens, therapeutic drugs, environmental toxins and products of oxidative stress, by conjugation with glutathione. The genes encoding the mu class of enzymes are organized in a gene cluster on chromosome 1p13.3 and are known to be highly polymorphic. These genetic variations can change an individual's susceptibility to carcinogens and toxins as well as affect the toxicity and efficacy of certain drugs. Diversification of these genes has occurred in regions encoding substrate-binding domains, as well as in tissue expression patterns, to accommodate an increasing number of foreign compounds.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTM5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GSTM5
Gene Accession No. P48774, Q9Z1B2
Gene Alias fibrous sheath component 2; Fsc2; glutathione S-alkyltransferase M5; glutathione S-aralkyltransferase M5; glutathione S-aryltransferase M5; glutathione S-transferase M5; glutathione S-transferase Mu 5; glutathione S-transferase, mu 5; glutathione-S-transferase, mu 5; GST class-mu 5; Gstm3; GSTM5; GSTM5-5; GTM5; S-(hydroxyalkyl)glutathione lyase M5
Gene Symbols GSTM5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human GSTM5 (NP_0008422).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14866, 64352
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.