Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTO1 Antibody (14A9.F6), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP244176AF700
Description
GSTO1 Monoclonal antibody specifically detects GSTO1 in Human, Mouse samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
GSTO1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
9446 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
14A9.F6 | |
Western Blot, ELISA, Immunohistochemistry | |
DKFZp686H13163, EC 2.5.1.18, glutathione S-transferase omega 1, glutathione S-transferase omega 1-1, glutathione S-transferase omega-1, glutathione-S-transferase like, GSTO-1, GSTTLp28, P28 | |
This Protein A purified GSTO1 Antibody (14A9.F6) was prepared by repeated immunizations in mice with GSTGSTO1 Protein (Uniprot: P78417) | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction