Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTO2 Antibody (OTI2A12), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272371V
Description
GSTO2 Monoclonal specifically detects GSTO2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
GSTO2 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GSTO2 | |
Full length recombinant protein of human GSTO2 (NP_899062) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2a |
Western Blot, Immunohistochemistry | |
OTI2A12 | |
Western Blot, Immunohistochemistry | |
bA127L20.1, bA127L20.1 (novel glutathione-S-transferase), EC 2.5.1.18, glutathione S-transferase omega 2, glutathione S-transferase omega-2, glutathione-S-transferase-like protein, GSTO-2 | |
Mouse | |
Affinity Purified | |
metabolism | |
119391.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction