Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTO2 Monoclonal Antibody (OTI2A12)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525814

Catalog No. PIMA525814


Only null left
Add to Cart

Description

Description

GSTO2 Monoclonal Antibody for Western Blot, IHC (P)

The omega class glutathione transferases (GST; EC 2.5.1.18) have poor activity with common GST substrates, but exhibit novel glutathione-dependent thioltransferase, dehydroascorbate reductase, and monomethylarsonate reductase activities, and they modulate Ca(2+) release by ryanodine receptors (e.g., RYR1).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GSTO2
Monoclonal
0.78 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H4Y5
Gsto2
Full length human recombinant protein of GSTO2 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI2A12
Unconjugated
Gsto2
1700020F09Rik; 4930425C18Rik; bA127L20.1; bA127L20.1 (novel glutathione-S-transferase); glutathione S-transferase omega 2; glutathione S-transferase omega 2-2; Glutathione S-transferase omega-2; glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase-like protein; GSTO 2-2; Gsto2; GSTO-2; LOC100720364; MMA(V) reductase; monomethylarsonic acid reductase
Mouse
Affinity Chromatography
RUO
119391
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.