Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTO2 Monoclonal Antibody (OTI4G1), TrueMAB™, OriGene
SDP

Catalog No. 502540765
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0765 30 μL
50-167-4516 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0765 Supplier OriGene Supplier No. TA503677S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GSTO2 Monoclonal antibody specifically detects GSTO2 in Human samples. It is validated for Western Blot

Specifications

Antigen GSTO2
Applications Western Blot
Classification Monoclonal
Clone OTI4G1
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DAND5
Gene Accession No. Q8N907
Gene Alias 1700020F09Rik; 4930425C18Rik; bA127L20.1; bA127L20.1 (novel glutathione-S-transferase); glutathione S-transferase omega 2; glutathione S-transferase omega 2-2; Glutathione S-transferase omega-2; glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase-like protein; GSTO 2-2; Gsto2; GSTO-2; LOC100720364; MMA(V) reductase; monomethylarsonic acid reductase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 23-189 of human DAND5 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 199699
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.