Learn More
GSTO2 Mouse anti-Human, Clone: OTI4H1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504085

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The omega class glutathione transferases (GST; EC 2.5.1.18) have poor activity with common GST substrates, but exhibit novel glutathione-dependent thioltransferase, dehydroascorbate reductase, and monomethylarsonate reductase activities, and they modulate Ca(2+) release by ryanodine receptors (e.g., RYR1).Specifications
GSTO2 | |
Monoclonal | |
Unconjugated | |
GSTO2 | |
GSTO 2-2, bA127L20.1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
119391 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4H1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H4Y5 | |
GSTO2 | |
Full length human recombit protein of human GSTO2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.