Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561664
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561664 Supplier Invitrogen™ Supplier No. PA561664
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVYGILDCVS HTPALRLWIS AMKWDPTVCA LLMDKSIFQG FLNLYFQNNP NAFDFG Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.

The omega class glutathione transferases (GST; EC 2.5.1.18) have poor activity with common GST substrates, but exhibit novel glutathione-dependent thioltransferase, dehydroascorbate reductase, and monomethylarsonate reductase activities, and they modulate Ca(2+) release by ryanodine receptors (e.g., RYR1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTO2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gsto2
Gene Accession No. Q9H4Y5
Gene Alias 1700020F09Rik; 4930425C18Rik; bA127L20.1; bA127L20.1 (novel glutathione-S-transferase); glutathione S-transferase omega 2; glutathione S-transferase omega 2-2; Glutathione S-transferase omega-2; glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase-like protein; GSTO 2-2; Gsto2; GSTO-2; LOC100720364; MMA(V) reductase; monomethylarsonic acid reductase
Gene Symbols Gsto2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GSTO2. Recombinant protein control fragment (Product #RP-100094).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119391
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.