Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596421 Supplier Invitrogen™ Supplier No. PA596421
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PHLTKECLVA LRCGRECTNL KAALRQEFSN LEEILEYQNT TFFGGTCISM IDYLLWPWFE RLDVYGILDC VSHTPALRLW ISAMKWDPTV CALLMDKSIF QGFLNLYFQN NPNAFDFGLC; Positive Samples: Rat testis, Mouse pancreas.

The omega class glutathione transferases (GST; EC 2.5.1.18) have poor activity with common GST substrates, but exhibit novel glutathione-dependent thioltransferase, dehydroascorbate reductase, and monomethylarsonate reductase activities, and they modulate Ca(2+) release by ryanodine receptors (e.g., RYR1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTO2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gsto2
Gene Accession No. Q6AXV9, Q8K2Q2
Gene Alias 1700020F09Rik; 4930425C18Rik; bA127L20.1; bA127L20.1 (novel glutathione-S-transferase); glutathione S-transferase omega 2; glutathione S-transferase omega 2-2; Glutathione S-transferase omega-2; glutathione-dependent dehydroascorbate reductase; glutathione-S-transferase-like protein; GSTO 2-2; Gsto2; GSTO-2; LOC100720364; MMA(V) reductase; monomethylarsonic acid reductase
Gene Symbols Gsto2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 124-243 of human GSTO2 (NP_8990621).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 309465, 68214
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.