Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTP1 Monoclonal Antibody (C2), Invitrogen™

Catalog No. FEMA551082
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551082 20 μL
FEMA551081 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551082 Supplier Invitrogen™ Supplier No. MA551082
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, the soluble GSTs are categorized into 4 main classes: alpha, mu, pi, and theta. This GST family member is a polymorphic gene encoding active, functionally different GSTP1 variant proteins that are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTP1
Applications Western Blot, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene Gstp1
Gene Accession No. P04906, P09211, P19157
Gene Alias Chain 7; deafness, X-linked 7; DFN7; epididymis secretory protein Li 22; FAEES3; fatty acid ethyl ester synthase III; glutathione S-transferase P; Glutathione S-transferase P 1; glutathione S-transferase pi 1; glutathione S-transferase pi 2; glutathione S-transferase, pi 1; glutathione S-transferase, pi 2; Glutathione-S-transferase placental enzyme pi type; glutathione-S-transferase, pi 1; Glutathione-S-transferase, placental enzyme pi type; GST 7-7; GST class-pi; gst P1; Gst p-1; GST YF-YF; Gst3; Gstp; GST-P; Gstp1; GSTP1-1; Gstp2; GstpiB; GST-piB; HEL-S-22; HGNC:4638; PI; Preadipocyte growth factor
Gene Symbols Gstp1
Host Species Mouse
Immunogen Recombinant Chinese hamster GSTp protein, Met1-Gln210 (Accession #P46424).
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100689311, 14870, 24426, 2950
Target Species Human, Mouse, Rat, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.