Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse Gstp2 His Protein

Click to view available options
Quantity:
0.02 mg
0.1 mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-210 Mouse of Mouse Gstp2 The Recombinant Mouse Gstp2 Protein is derived from E. coli. The Recombinant Mouse Gstp2 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 1mg/mL |
For Use With (Application) | SDS-PAGE |
Formulation | Phosphate buffered saline (pH 7.4) containing 10% glycerol |
Molecular Weight (g/mol) | TMW: 25.9kDa |
Quantity | 0.1 mg |
Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
Common Name | Gstp2 |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction