Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva GSTrap™ FF Columns: 7 x 25mm

Catalog No. p-4892091
Click to view available options
Quantity:
2 x 1 mL
5 x 1 mL

Purifies GST-tagged proteins in one step

  • Mild elution conditions preserve antigenicity and function of proteins extracted
  • Prepacked with Glutathione Sepharose™ media
  • Average particle size: 90μm
  • Bed dimensions: 7 × 25mm
  • Flow velocity: <4mL/min.
Media
  • Matrix: Highly cross-linked 4% agarose
  • Ligand: glutathione and 10-carbon linker arm
  • Binding capacity: >10mg/mL
  • pH stability working range: 3 to 12

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 1 mL
Diameter (English) Inner 0.27 in.
Diameter (Metric) Inner 7 mm
Description GSTrap FF columns are prepacked Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins.
Flow Rate <4 mL/min.
Material Polypropylene
pH Range 3 to 12
Support Type 4% Highly Cross-linked Agarose
Content And Storage 4°C to 30°C, 20% Ethanol
Max. Operating Pressure 5 bar (0.5 MPa, 70 psi)
Particle Size 90 μm
Quantity 5 x 1 mL
For Use With (Application) For the purification of GST-tagged proteins from bacterial lysates and other glutathione S-tranferases or glutathione-dependent proteins and for preparation of cytoplasmic extract
Type Pre-packed Column
Ligand Type Glutathione and 10 carbon linker arm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.