Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva GSTrap™ HP Columns

Catalog No. 45000333 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5 mL
5 x 1 mL
5 x 5 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-333 1 x 5 mL
45-000-332 5 x 1 mL
45-000-334 5 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 45-000-333 Supplier Cytiva Supplier No. 17528201
Add to Cart
Add to Cart
This item is not returnable. View return policy

Use to provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. Cytiva Life Sciences™ GSTrap™ HP Columns are prepacked with Glutathione Sepharose™ High Performance medium.

  • One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
  • Glutathione coupled to Sepharose™ High Performance media
  • Column material: polypropylene
  • May be used with syringe, peristaltic pump or chromatography system
  • Purification process preserves antigenicity and function
  • Available as 1mL and 5mL columns
Media
  • Average particle size: 34μm
  • Ligand: Glutathione
  • Matrix: Highly cross-linked 6% agarose
  • Binding capacity: >7mg/mL
  • pH stability working range: 3 to 12

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.
5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 5 mL
Diameter (English) Inner 0.62 in.
Diameter (Metric) Inner 16 mm
Description GSTrap HP columns are prepacked with Glutathione Sepharose High Performance resin and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins.
Flow Rate <20 mL/min.
Material Polypropylene
pH Range 3 to 12
Support Type 4% Highly Cross-linked Agarose
Content And Storage 4°C to 30°C, 20% Ethanol
Max. Operating Pressure 5 bar (0.5 MPa, 70 psi)
Particle Size 34 μm
Quantity 1 x 5 mL
For Use With (Application) Glutathione S-transferase (GST) tagged proteins produced using the pGEX series of expression vectors, Other glutathione S-transferases and glutathione-dependent proteins
Type Pre-packed Column
Ligand Type Glutathione
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.